missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBZ Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19995-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to HBZ.
Zeta-globin is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that includes five functional genes and two pseudogenes. The order of genes is: 5′ - zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 -alpha-1 - theta1 - 3′. Zeta-globin (HBZ ) is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that involves 4 functional genes and 3 nonfunctional pseudogenes. The order of genes is: 5′-zeta -- pseudozeta -- pseudoalpha2 -- pseudoalpha1 -- alpha2 -- alph1 -- theta1-3′.Specifications
HBZ | |
Polyclonal | |
Unconjugated | |
HBZ | |
HBAP2, HBAZ, HBM, HBZ, HBZ2, Hemoglobin mu chain, Hemoglobin subunit mu, Hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, mu, hemoglobin, zeta, Mu globin, Zeta globin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3050 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02008 | |
HBZ | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |