Learn More
Gibco™ HBSS, calcium, magnesium
GREENER_CHOICE
Product Code. 11580616
Change view
Click to view available options
Quantity:
100 mL
500 mL
10 x 500 mL
20 x 100 mL
Unit Size:
1 set
100mL
2000mL
500mL
Product Code. Quantity unitSize
11580616 20 x 100 mL 2000mL
11550616 100 mL 100mL
11560616 500 mL 500mL
11570616 10 x 500 mL 1 set
4 options
Product Code. 11580616
Brand: Gibco™ 24020141
HBSS, calcium, magnesium
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility (24020-117). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 15 to 30°C
Description Hanks' Balanced Salt Solution, 20 x 100mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glucose, Calcium, Magnesium, Sodium Bicarbonate, Phenol Red, 36 Months Shelf Life, For Mammalian Cell Culture
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
With Additives Glucose, Calcium, Magnesium, Sodium Bicarbonate, Phenol Red
For Use With (Application) Mammalian Cell Culture
pH 7.0 to 7.4
Quantity 20 x 100 mL
Regulatory Status For in-vitro diagnostic use