missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (HBsAg/7556R), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24242AF594
5236.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HBsAg | |
| Monoclonal | |
| Alexa Fluor 594 | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Recombinant full-length human HBsAg protein | |
| 0.1 mL | |
| Cell Biology, Virology Bacteria and Parasites | |
| 944569 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| HBsAg/7556R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction