Learn More
Horst™ HBS Model Smooth Heating Tape
Product Code. p-8005781
Click to view available options
Dimensions (L x W):
0.5 Lm
1 Lm
1.5 Lm
10 Lm
2 Lm
2.5 Lm
3 Lm
4 Lm
5 Lm
Unit Size:
Each
Product Code. 10608472
Brand: Horst™ 20301
Only null left
Add to basket
Add to basket
This versatile heating tape meets high requirements regarding heating power and operating temperature in a dry environment.
  • Nominal voltage: 230 V∼
  • Connection cable: 0.5 m
  • Thickness: 5.5 mm
  • Breadth: 30 mm
  • Min. bending radius: 10 mm
  • Protective braid: Nickel
Specifications
Dimensions (L x W) 0.5 Lm
Dimensions (L x W x H) 0.5 Lm
Electrical Requirements ∼230V Voltage
Model HBS
Product Type Smooth Heating Tape