Learn More
HBQ1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16706-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16896424
  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
Description
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5′ - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3′. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue.
Specifications
Specifications
HBQ1
Polyclonal
Unconjugated
HBQ1
HBQ1, Hemoglobin subunit theta 1, Hemoglobin theta 1 chain, hemoglobin, theta 1, Theta 1 globin
Rabbit
Antigen Affinity Chromatography
RUO
3049
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09105
HBQ1
HBQ1 Fusion Protein Ag10105