missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HBG1 Recombinant Protein
Marque: Abnova™ H00003047-P01.10ug
Informations supplémentaires : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
AAH10913 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
41.9 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MGHFTEEDKATITSLWGKVNVEDAGGETLGRLLVVYPWTQRFFDSFGNLSSASAVMGNPKVKAHGKKVLTSLGDAIKHLDDLKGTFAQLSELHCDKLHVDPENFKLLGNVLVTVLAIHFGKEFTPEVQASWQKMVTGVASALSSRYH | |
HBGA/HBGR/HSGGL1/PRO2979 | |
HBG1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
3047 | |
HBG1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBG1 | |
Human | |
Recombinant | |
Solution |