Learn More
Horst™ HB Heating Tape
Product Code. 10097021
Click to view available options
Length (Metric):
1.0 m
1.5 m
2.0 m
Wattage:
250w
350w
500w
Unit Size:
Each
Product Code. 10097021
Brand: Horst™ 20203
Specifications
Length (Metric) 1.5 m
Voltage 230 V
Model HB
Length (Metric) Power Cord 0.5 m
Wattage 350w
Width (Metric) 30mm
Product Type Heating Tape
For Use With (Application) For Very Small Winding Radii