Learn More
3M™ Hazardous Spill Response Kit
Product Code. p-6838007
Click to view available options
Capacity (Metric):
5 L
26 L
Dimensions (L x W):
400 x 70 mm
500 x 630 mm
Includes:
10 sheets, 1 pillow, 1 disposable bag with tie
10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties.
Unit Size:
Each
Product Code. 12327979
Brand: 3M™ 7100090073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Contains chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. 3M™ Hazardous Spill Response Kits can be used to contain and clean up the hazardous liquid without delay.
Kits contain chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. In the event of a spill, the kit can be used to contain and clean up the hazardous liquid without delay.
  • Constructed from polypropylene fibers
  • Available in a wide range of formats
  • Lightweight and dust-free
  • High absorption capacity minimizes waste for disposal.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 26 L
Material Polypropylene Fiber
Type Spill Kit
Includes 10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties.
Packaging Quantity 1 Ea.
Dimensions (L x W) 500 x 630 mm