Learn More
3M™ Hazard Warning Tape
Product Code. 17267790
Change view
Click to view available options
Color:
Black with Yellow Stripes
Red
Unit Size:
24 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17267790 Black with Yellow Stripes 24 rolls
17277790 Red 24 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17267790 Supplier 3M™ Supplier No. 7100015263
  • ONLINE EXCLUSIVE PRICE
    4475.00 NOK
    2847.70 NOK / 24 rolls
    Save 1627.30 NOK
    36% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Used for marking hazardous areas, protruding equipment, or preventing accidents in factories, warehouses and hospitals.
  • Use for taping in all non-critical applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas
TRUSTED_SUSTAINABILITY
Specifications
Description 766 i
Dimensions (L x W) 50 mm x 33 m
Length (Metric) 33 m
Width (Metric) 50 mm
Color Black with Yellow Stripes