Learn More
HAUS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18991-1-AP-20UL
This item is not returnable.
View return policy
Description
Contributes to mitotic spindle assembly, maintece of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.Specifications
| HAUS3 | |
| Polyclonal | |
| Unconjugated | |
| HAUS3 | |
| C4orf15, HAUS3, IT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231123, 680477, 79441 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q68CZ6, Q8QZX2 | |
| HAUS3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |