Learn More
Brady™ Harsh Environment Multipurpose Polyester Labels for M710 and BMP71
Click to view available options
Dimensions (LxW):
12.7 x 25.4 mm
25.4 x 25.4 mm
4.75 x 25.4 mm
6.35 x 22.86 mm
6.99 x 25.4 mm
9.53 x 25.4 mm
Shape:
Rectangle
Square
missing translation for 'unitSize'
Pack of 250
Pack of 500
Pack of 750
Description
- Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification
- Glossy white finish with permanent acrylic is thermal transfer printable
- Meets requirements of a halogen-free material per DIN VDE 0472 part 815
- Service temperature of -70°C to +110°C (-94°F to +230°F)
- Brady™ authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability
- A ribbon is required to print this material
Specifications
Specifications
| Certifications/Compliance | Halogen-free (DIN VDE 0472 Part 815) |
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Color | White |
| Finish | Gloss |
| Format | Label |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Show More |