Learn More
Hardwood Test Tube Racks
Click to view available options
Positions:
2 and 10
Unit Size:
Pack of 5
Description
Single row, can be packed flat, comprising hardwood top and bottom members with moulded polypropylene supports which clip into position. Occupy only about one third of the space required by a conventional rack when dismantled for storage.
Specifications
Specifications
| Product Type | Test Tube Rack |
| Holds | Tubes (22 and 32 mm) |
| Material | Hardwood |
| Positions | 2 and 10 |
| Diameter (Metric) Opening | 32 mm and 22 mm |