Learn More
Hardwood Combined Funnel/Burette Stand
Product Code. 12946796
Change view
Click to view available options
Dimensions (L x W):
290 x 100 mm (Base)
Unit Size:
Each
Product Code. Dimensions (L x W) unitSize
12946796 290 x 100 mm (Base) Each
1 options
Product Code. 12946796
Brand: S Murray SAMCO L328/01
Only null left
Add to basket
Add to basket
Hardwood, with interchangeable components
Hardwood, with interchangeable components. Bases are 205mm x 110mm (single) and 290mm x 110mm (double). Rods are 460mm x 16mm (dia). Burette clamps have lined jaws and funnel holders are slotted; all are fitted with a plastic screw and rubber feet on base.
Specifications
Material Hardwood
Dimensions (L x W) 290 x 100 mm (Base)
Length (Metric) Rod 460 mm
Diameter (Metric) Rod 16 mm
Product Type Combined Funnel/Burette Stand
Quantity 1 Ea.