Learn More
Sartorius Hardware Package for Built-in Ionizer
Product Code. 17987320 Shop All Sartorius Products
Change view
Click to view available options
:
Type: Software License
Product Code.
17987320 Type: Software License
1 options
Product Code. 17987320
Brand: Sartorius QAPP1001
Activation Ionizer accessory for Cubis™ II for effectively neutralizing charges from samples, containers, or powders.
  • Built-in Ionizer Upgrade for Sartorius Cubis II Ultra Hi-Resolution High-Capacity Micro Balances
  • Effectively neutralizes electrostatic charges from samples, container, or powder
  • Helps ensure accurate results and stable readouts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cubis™ II High-Capacity Micro Balance
Type Software License