Learn More
Sartorius Hardware Package for Automatic Draft Shield
Product Code. 17937330 Shop All Sartorius Products
Change view
Click to view available options
:
Type: Software License
Product Code.
17937330 Type: Software License
1 options
Product Code. 17937330
Brand: Sartorius QAPP1002
Activation of Motorized Draft Shield for Sartorius Cubis™ II ultra hi-resolution high-capacity micro balances.
  • Activation of Motorized Draft Shield for Sartorius Cubis™ II Ultra Hi-Resolution High-Capacity Micro Balances
  • Removable and easy-to-clean; designed for tool-free disassembly
  • Material: glass
  • Configurable on models with MCA or MCE Interface at time of purchase
  • After purchase licensing only available for MCA models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cubis™ II High-Capacity Micro Balance
Type Software License