Learn More
Nabertherm™ Hardening Furnace
Product Code. p-7236248 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
9 L
11 L
17 L
30 L
40 L
60 L
Controller Type:
B500
C540
Temperature Control:
Minutes to Max Temp: 105
Minutes to Max Temp: 120
Minutes to Max Temp: 180
Unit Size:
Each
Product Code. Capacity (Metric) Controller Type Temperature Control unitSize
17308029 9 L C540 Minutes to Max Temp: 180 Each
17318029 11 L B500 Minutes to Max Temp: 180 Each
17328029 11 L C540 Minutes to Max Temp: 180 Each
17338029 11 L B500 Minutes to Max Temp: 120 Each
17348029 11 L C540 Minutes to Max Temp: 120 Each
17358029 17 L B500 Minutes to Max Temp: 120 Each
17368029 17 L C540 Minutes to Max Temp: 120 Each
17378029 30 L B500 Minutes to Max Temp: 105 Each
17398029 40 L B500 Minutes to Max Temp: 120 Each
17308039 40 L C540 Minutes to Max Temp: 120 Each
17318039 60 L B500 Minutes to Max Temp: 120 Each
11 options
Product Code. 17308029
Brand: Nabertherm™ N01H1NMNA
Only null left
Add to basket
Add to basket
The chamber furnaces are a perfect fit to withstand harsh use in the laboratory, e.g. when heat-treating metals, robust insulation with light refractory bricks is necessary. The furnaces can be extended with a variety of accessories.
  • Max Temperature: 1280°C
Specifications
Controller Type C540
Capacity (Metric) 9 L
Temperature (Metric) 1280°C
Type Hardening Furnace
Style Tabletop
Temperature Control Minutes to Max Temp: 180
Dimensions (D x W x H) Interior 250 x 250 x 140 mm
Dimensions (L x W x H) Exterior 800 x 650 x 600 mm