Description
- Max Temperature: 1280°C
Specifications
Specifications
| Controller Type | B500 |
| Capacity (Metric) | 60 L |
| Temperature (Metric) | 1280°C |
| Type | Hardening Furnace |
| Temperature Control | Minutes to Max Temp: 120 |
| Dimensions (D x W x H) Interior | 350 x 750 x 250 mm |
| Dimensions (L x W x H) Exterior | 1040 x 1500 x 1340 mm |