Learn More
Brady™ Ultimate Security Padlock
Click to view available options
Height (Metric) Shackle:
26.00 mm
50.00 mm
75.00 mm
Unit Size:
Each
Pack of 3
Description
- Shackle and body are made of highest standard hardened alloy steel
Specifications
Specifications
| Product Type | Ultimate Security Padlock |
| Color | Black |
| Material | Hardened Alloy Steel |
| Description | Ultimate Security Padlock |
| Diameter (Metric) Shackle | 11 mm |
| Dimensions (L x W) | 63.00 x 85.00 mm |
| Height (Metric) | 85 mm |
| Height (Metric) Shackle | 26.00 mm |
| Material (Shackle) | Hardened Alloy Steel |
| No. of Pins | 7 |
| Show More |