Learn More
Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop
Click to view available options
Packaging Type:
Bag
Singles in peel pouch
Color:
Clear
Green
Unit Size:
Pack of 1000
Description
- Cadmium-free pigments
- Gamma Irradiation to achieve a SAL of 10-6
- Shelf Life: 3 years
- 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Material | Hard Crystal Polystyrene |
| Sterility | Sterile |
| Color | Clear |
| Product Type | Inoculating Loop |
| Quantity | 50/bag |
| Packaging Type | Bag |