missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haptoglobin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16665-1-AP-20UL
This item is not returnable.
View return policy
Description
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which aslo acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chanins bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.Specifications
| Haptoglobin | |
| Polyclonal | |
| Unconjugated | |
| HP | |
| BP, HAPT, Haptoglobin, HP, HP2 ALPHA 2, HPA1S | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15439, 24464, 3240 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00738, P06866, Q61646 | |
| Hp | |
| Haptoglobin Fusion Protein Ag10143 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction