Learn More
Plum™ Handy Plus Skin Lotion
Product Code. 15751137
Change view
Click to view available options
Packaging:
8 x 0.7 L bag-in-box
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
15751137 8 x 0.7 L bag-in-box Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15751137 Supplier Plum™ Supplier No. P2903
  • ONLINE EXCLUSIVE PRICE
    1800.00 NOK
    1410.44 NOK / Pack of 8
    Save 389.56 NOK
    21% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Mildly perfumed, replenishing skin care lotion for normal and dry skin. Well-suited for offices, administrative areas and public toilets.
  • Replenishing: contributes to maintaining the natural barrier properties of the skin
  • Mildly perfumed, though colour and paraben-free
  • Quickly absorbed by the skin and contributes to the skin moisture properties of the skin
  • Specifications
    Product Type Skin Care Lotion
    Packaging 8 x 0.7 L bag-in-box
    Quantity 8 Pk.
    Volume (Metric) 0.7 L