Learn More
Sampling Systems™ Handle Extension Rod for Dipper
Product Code. 15889361 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
600 mm
Unit Size:
Each
Product Code. 15889361
Brand: Sampling Systems™ 5100E600
Only null left
Add to basket
Add to basket
Ideal for obtaining surface samples from a wide range of liquids. Sampling Systems™ Handle Extension Rod for PTFE Dippers are quick and easy to use.
  • 600mm long removable handle extension.
  • For use with 5100A-100, 5100A-250, 5100A-500 and 5100A-1000
  • Not suitable for use with 5100A-10 which has a 155mm long fixed handle
Specifications
Material PTFE
For Use With (Equipment) PTFE Dipper
Length (Metric) 600 mm