Learn More
Brady™ HandiMark™ Vinyl Repositionable Film
Specifications
Specifications
| Description | HandiMark Repositionable Vinyl Film |
| Adhesive Type | Acrylic, Repositionable |
| Color | Yellow |
| Finish | Gloss |
| Material | Vinyl |
| Product Type | Repositionable Film |
| For Use With (Application) | For temporary, removable and repositionable labels, like 5S and lean inventory labels |
| For Use With (Equipment) | HandiMark |
| Quantity | 1 Roll |
| Temperature Range (Metric) | -40°C to +82°C |
| Show More |