Learn More
Buerkle™ Hand Scoops for Food stuffs
Product Code. 31188869 Shop All Bürkle Products
Change view
Click to view available options
Volume (Metric):
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
31188869 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31188869 Supplier Buerkle™ Supplier No. 83001010
Estimated Shipment: 16-10-2026
Polypropylene's easy-to-clean, smooth and pore-free surface makes these scoops extremely suitable for the food sector. The scoops can be also used for boiling products.
Iron scoops are loud, they can be dented, they rust and they are heavy. Plastic scoops have none of these disadvantages and are also hygienic because they are made in one piece and have a non-stick surface. They also have a high chemical resistance, they are unbreakable and all valuable advantages!
  • For foodstuffs (FDA and (EU) Nr. 10/2011)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scoop
Material Polypropylene
Volume (Metric) 2 L