Learn More
Kimberly-Clark™ Scott™ Luxury Foam Frequent Use Hand Cleanser
Product Code. 12631376
Change view
Click to view available options
Volume (Metric):
1000 mL
1200 mL
Unit Size:
Pack of 4
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12631376 1000 mL Pack of 6
12621356 1200 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12631376 Supplier Kimberly-Clark™ Supplier No. 6342
  • ONLINE EXCLUSIVE PRICE
    1620.00 NOK
    527.01 NOK / Pack of 6
    Save 1092.99 NOK
    67% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Dispenses the ideal amount of soap for each wash preventing overuse. Scott™ Luxury Foam Frequent Use Hand Cleanser delivers up to twice as many washes per liter as conventional liquid soaps.
  • Color-free and fragrance-free foam
  • Luxuriant feel formulated for frequent use
  • Strong, hygienically sealed cassettes that are quick and easy to install
  • Give up to 2,500 shots of soap
  • Cassettes shrink to save space when empty and are recyclable (after the pump is removed)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Cleanser
Color Clear
Certifications/Compliance HACCP Certification, FSC
Quantity 6 Cassettes
Size Up to 2,500 shots
Volume (English) 33.8 oz.
Volume (Metric) 1000 mL