Learn More
Hammond™ Drierite™ Desiccant Bags, Cole-Parmer™
Product Code. 11803092 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
3 g
6 g
1 oz.
8 oz.
Unit Size:
Pack of 100
Pack of 200
Pack of 750
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11803092 1 oz. Pack of 200
11832692 3 g Pack of 750
11893082 6 g Pack of 750
11833092 8 oz. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11803092 Supplier Cole Parmer Supplier No. 0719308
  • ONLINE EXCLUSIVE PRICE
    2360.00 NOK
    1125.56 NOK / Pack of 200
    Save 1234.44 NOK
    52% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Protect materials from corrosion with these desiccant bags. Hammond™ Drierite™ Desiccant Bags reduce the humidity to a 100°F dew point in 10 hours and are dust-tight. Materials packaged with these bags are protected from corrosion for many months.
Nonwoven crepe bags filled with Drierite (anyhydrous calcium sulfate) completely dry the air within a package or container during shipment or storage.
  • Sturdy and held together by heat seals
  • Perform well in elevated temperatures
  • Will not release absorbed moisture at less than 350°F
Specifications
Material Calcium Sulfate
Quantity 1 oz.
Suitability These Bags are Dust-Tight and Will Protect For Many Months
compliance-icons
Product Identifier
  • BAG DRIERITE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: calcium sulphate, cobalt chloride