Learn More
Hamilton™ Stainless Steel Guard Column Holder
Product Code. 10025360
Change view
Click to view available options
Pore Size:
2 μm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
10025360 2 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10025360 Supplier Hamilton™ Supplier No. 32908
  • ONLINE EXCLUSIVE PRICE
    2205.00 NOK
    1942.52 NOK / Pack of 1
    Save 262.48 NOK
    11% Off
Estimated Shipment: 26-10-2026
Stainless steel and PEEK guard columns help protect your investment
  • Protects analytical HPLC columns
  • Removes particulate contaminants and highly adsorptive compounds from samples, prolonging analytical column life
  • Modular and cartridge replacement is easy and tool free
  • Available in stainless steel and PEEK materials, optimized to reduce dead volume and prevent extra column band broadening
  • Holders are indefinitely reusable
  • Guard Columns for Stainless Steel HPLC Columns: Recommended for use with virtually all solvents except high concentrations of sodium chloride
  • Guard Columns for PEEK HPLC Columns: Recommended for use with virtually all solvents except, concentrated nitric acid, sulfuric acid, tetrahydrofuran or dimethylsulfoxide
Specifications
Diameter (Metric) Inner 2.3 mm
Length (Metric) 25 mm
Particle Size 12 to 20μm Polymeric Packing, 10μm for Silica Packing
Pore Size 2 μm
For Use With (Equipment) Virtually all Solvents except High Concentrations of Sodium Chloride
Product Type Holder Assembly