Learn More
- Home
- Products
- Chromatography Syringes
- Chromatography Syringe Accessories
- Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)
Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)
Product Code. 10225142
Change view
Click to view available options
Needle Gauge:
22 G
22s G
26 G
26s G
27 G
28 G
30 G
31 G
32 G
33 G
Needle Point Style:
Blunt
Cone
Sharp, Bevel, Curved
Diameter (English) Inner:
0.004 in.
0.005 in.
0.0051 in.
0.0059 in.
0.0063 in.
0.007 in.
0.01 in.
0.016 in.
Diameter (English) Outer:
0.008 in.
0.0094 in.
0.01 in.
0.012 in.
0.014 in.
0.016 in.
0.018 in.
0.019 in.
0.028 in.
Unit Size:
Pack of 6
| Product Code. | Needle Gauge | Needle Point Style | Diameter (English) Inner | Diameter (English) Outer | unitSize |
|---|---|---|---|---|---|
| 10225142 | 26s G | Blunt | 0.0051 in. | 0.018 in. | Pack of 6 |
| 10291733 | 26s G | Cone | 0.0051 in. | 0.018 in. | Pack of 6 |
| 10708494 | 26 G | Cone | 0.01 in. | 0.018 in. | Pack of 6 |
| 10284302 | 22 G | Cone | 0.016 in. | 0.028 in. | Pack of 6 |
| 10622804 | 22 G | Sharp, Bevel, Curved | 0.016 in. | 0.028 in. | Pack of 6 |
| 10274702 | 22s G | Cone | 0.0059 in. | 0.028 in. | Pack of 6 |
| 10323811 | 22s G | Sharp, Bevel, Curved | 0.0059 in. | 0.028 in. | Pack of 6 |
| 11557704 | 22s G | Blunt | 0.0059 in. | 0.028 in. | Pack of 6 |
| 11520142 | 33 G | Blunt | 0.004 in. | 0.008 in. | Pack of 6 |
| 10152642 | 22 G | Blunt | 0.016 in. | 0.028 in. | Pack of 6 |
| 10788731 | 26s G | Sharp, Bevel, Curved | 0.0051 in. | 0.019 in. | Pack of 6 |
| 11547704 | 26 G | Blunt | 0.01 in. | 0.01 in. | Pack of 6 |
| 11503234 | 26 G | Sharp, Bevel, Curved | 0.01 in. | 0.018 in. | Pack of 6 |
| 10616264 | 30 G | Blunt | 0.0063 in. | 0.012 in. | Pack of 6 |
| 11510142 | 32 G | Blunt | 0.004 in. | 0.0094 in. | Pack of 6 |
| 10183352 | 31 G | Blunt | 0.005 in. | 0.01 in. | Pack of 6 |
| 11500142 | 28 G | Blunt | 0.007 in. | 0.014 in. | Pack of 6 |
| 10281293 | 27 G | Blunt | 0.0051 in. | 0.016 in. | Pack of 6 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Standard needle length is 51mm (2 in.)
- Custom gauges, lengths and point styles are available
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel (Hub) |
| Needle Gauge | 26s G |
| Diameter (English) Inner | 0.0051 in. |
| Diameter (English) Outer | 0.018 in. |
| Diameter (Metric) Inner | 0.13 mm |
| Diameter (Metric) Outer | 0.47 mm |
| For Use With (Application) | Minimises risk of needle clogging |
| Length (English) Needle | 2 in. |
| Product Type | Replacement Syringe Needle |
| Needle Point Style | Blunt |
| Show More Show Less |