Learn More
Hamilton™ PTFE Tubing Adapted to Femaile Luer Lock
Description
- Made of PTFE
- One female luer fitting on the end
- Tubing is 12 in. in length
Specifications
Specifications
| Type | Tubing Assembly |
| Length (English) | 12 in. |
| Material | PTFE |
| Length (Metric) | 30.48 cm |
| For Use With (Application) | Connect to a syringe or other component |
| Needle Gauge | 18 G |