Learn More
Hamilton™ PTFE Tubing Adapted to Femaile Luer Lock
Click to view available options
Needle Gauge:
15 G
18 G
19 G
20 G
22 G
24 G
28 G
30 G
Unit Size:
Each
Description
- Made of PTFE
- One female luer fitting on the end
- Tubing is 12 in. in length
Specifications
Specifications
| Type | Tubing Assembly |
| Length (English) | 12 in. |
| Material | PTFE |
| Length (Metric) | 30.48 cm |
| For Use With (Application) | Connect to a syringe or other component |
| Needle Gauge | 30 G |