Learn More
Hamilton™ PRP™-X100 Guard Cartridge Kit
Product Code. 10220773
Change view
Click to view available options
Material:
PEEK
Stainless Steel
Column Capacity:
57 μL
80 μL
Length (Metric):
8 mm
20 mm
missing translation for 'unitSize'
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Column Capacity Length (Metric) unitSize
10220773 Stainless Steel 80 μL 20 mm Each
10073204 PEEK 57 μL 8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 10220773 missing translation for 'mfr' Hamilton™ missing translation for 'supplierNo' 79448
Only null left
Add to basket
Add to basket
Stainless steel and PEEK guard columns help protect your investment
  • Guard columns remove particulate contaminants and highly adsorptive compounds from samples
  • Modular and cartridge replacement is easy and tool free
  • Holders are available in stainless steel and PEEK materials, both designs optimized to reduce dead volume and prevent extra column band broadening
  • Holders for the cartridges are reusable indefinitely
Kit contains 1 holder and 2 cartridges
Recommended for use with virtually all solvents except high concentrations of Sodium Chloride.
Specifications
Particle Size 10 μm
Length (Metric) 20 mm
Max. Pressure 5000 psi
Column Capacity 80 μL
Diameter (Metric) Inner 2.3 mm
Material Stainless Steel
For Use With (Application) Virtually all solvents except high concentrations of sodium chloride