Learn More
Hamilton™ Neuros Technology Adapter Kits for 700/1700 Series Neuros Syringes
Product Code. 15142654
Change view
Click to view available options
Needle Gauge:
33 G
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
15142654 33 G Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15142654 Supplier Hamilton™ Supplier No. 6546001
  • ONLINE EXCLUSIVE PRICE
    4120.00 NOK
    3758.35 NOK / Pack of 1
    Save 361.65 NOK
    8% Off
Estimated Shipment: 19-11-2026
Includes
Kit includes adapter, two sleeves, 6-pack of 33 gauge needles
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Order Info
  • This adapter kit is for Hamilton 700 and 1700 Series Syringes only. The Hamilton 7000 Series Neuros Syringes come equipped with Neuros technology and do not require an adapter kit.
  • Replacement needles are available in six packs (Fisher P/N: 14-815-905, Vendor P/N: 65461-01)
Specifications
Needle Gauge 33 G
Includes 1 borosilicate glass adapter, 2 sleeves, 6 pack of 33G needles
Product Type Adapter Kit
For Use With (Equipment) Neuros 700/1700 Series Microliter or Gastight Syringes