Learn More
Hamilton™ Neuros Replacement Needles for Microliter/Gastight™ Syringes
Product Code. 13400899
Change view
Click to view available options
Needle Gauge:
33 G
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Unit Size
13400899 33 G Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13400899 Supplier Hamilton™ Supplier No. 6546101
  • ONLINE EXCLUSIVE PRICE
    2750.00 NOK
    2412.87 NOK / Pack of 6
    Save 337.13 NOK
    12% Off
Estimated Shipment: 11-12-2026
Removable needles can be used with select Hamilton Removable Needle (RN) 700 and 1700 Series Neuros syringes
  • Special blunt needle point suited for neuroscience applications
  • 304 stainless steel
  • 33 gauge
  • Blunt point (Point Style 3)
  • Exposed needle length can be adjusted by the user from 0 to 20mm
  • Sold in 6-packs
Neuros replacement needles work with the following Hamilton 700 and 1700 Series syringes:
  • Model 75 RN Syringe (14-824-651)
  • Model 1701 RN Syringe (14-824-652)
  • Model 1702 RN Syringe (14-824-653)
  • Model 1705 RN Syringe (14-824-654)
  • Model 1710 RN Syringe (14-824-655)
Specifications
Autoclavable Autoclavable
Needle Gauge 33 G
Diameter (English) Inner 0.00015 in.
Diameter (English) Outer 0.0003 in.
Diameter (Metric) Inner 0.004 mm
Diameter (Metric) Outer 0.008 mm
For Use With (Application) Used for neuroscience applications and select Hamilton removable needle 700 and 1700 series 0.79μL neuros syringes
Product Type Replacement Needle
Needle Point Style Removable Square/Blunt