Learn More
Hamilton™ Tungsten Cleaning Wires
Description
Tungsten wires, best used with Hamilton Company Needle Cleaning Concentrate (14-819-85, 14-819-86).
Order Info
Ten wires per pack.
Specifications
Specifications
| Material | Tungsten |
| Length (English) | 7 in. |
| Length (Metric) | 17.78 cm |
| Product Type | Cleaning Wire |
| For Use With (Equipment) | 24 to 26G Needle, Syringe and Needle Cleaning Kit |