Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
Product Code. 11684133
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Model:
1001 BFP
1002 BFP
1005 BFP
1010 BFP
1701 BFP
1702 BFP
1705 BFP
1710 BFP
1725 BFP
1750 BFP
Bubble free prime
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Model Unit Size
11684133 10 mL 1010 BFP Each
15142644 10 μL 1701 BFP Each
15122644 50 μL 1705 BFP Each
12638473 100 μL 1710 BFP Each
15132644 25 μL 1702 BFP Each
15146862 25 μL 1702 BFP Each
12396239 250 μL 1725 BFP Each
13478496 500 μL 1750 BFP Each
12528126 1 mL 1001 BFP Each
13346319 2.5 mL 1002 BFP Each
15182644 50 mL Bubble free prime Each
15192644 25 mL Bubble free prime Each
12316249 5 mL 1005 BFP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11684133 Supplier Hamilton™ Supplier No. 5900050
  • ONLINE EXCLUSIVE PRICE
    3140.00 NOK
    2819.84 NOK / Pack of 1
    Save 320.16 NOK
    10% Off
Estimated Shipment: 09-12-2026
For use with MICROLAB 600 Diluter/Dispenser
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Material Borosilicate Glass
Needle Gauge No Needle Included
Product Type Gastight Syringe
Model 1010 BFP
Volume (Metric) 10 mL
For Use With (Equipment) Microlab™ 600 dispensers
Connection Type Bubble Free Prime
Graduated Yes