Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Product Code. 15301530
Change view
Click to view available options
Needle Gauge:
14 G
17 G
18 G
19 G
20 G
21 G
22 G
22s G
23 G
24 G
25 G
26 G
26s G
33 G
Unit Size:
Pack of 6
Product Code. Needle Gauge unitSize
15301530 19 G Pack of 6
11557794 26 G Pack of 6
11592315 14 G Pack of 6
11597784 17 G Pack of 6
11507794 18 G Pack of 6
11517794 20 G Pack of 6
11527794 21 G Pack of 6
11537794 23 G Pack of 6
10422172 22 G Pack of 6
11547794 24 G Pack of 6
10775944 25 G Pack of 6
11572325 33 G Pack of 6
11587794 26s G Pack of 6
10587833 22s G Pack of 6
14 options
Product Code. 15301530
Brand: Hamilton™ 91019
Can be used with TLL syringes and LT or TLL connectors.
  • Made of 304 Stainless Steel
  • Standard needle length is 51mm (2 in.)
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
  • Needles are available from 10 to 33 ga
Specifications
Autoclavable Autoclavable
Material Nickel-plated Brass (Hub), Stainless Steel
Needle Gauge 19 G
Diameter (English) Inner 0.027 in.
Diameter (English) Outer 0.042 in.
Diameter (Metric) Inner 0.69 mm
Diameter (Metric) Outer 1 mm
Length (English) Needle 2 in.
Product Type Needle
Needle Point Style Blunt
For Use With (Equipment) PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors