Learn More
Hamilton™ Gastight™ Waters WISP Autosampler Syringes
Product Code. 11520222
Click to view available options
Volume (Metric):
25 μL
250 μL
Max. Pressure:
1000 psi
500 psi
Model:
1702C
1725C
Unit Size:
Each
Product Code. 11520222
Brand: Hamilton™ 80020
Designed for use with Waters WISP HPLC autosamplers
Tight fit plunger tip wipes the syringe barrel interior free of sample. This feature is useful with heterogeneous samples as it reduces chances that a deposit will occur and cause plunger to freeze.
  • For dispensing both liquids and gases
  • Precision-machined PTFE plunger tip creates leak-free seal; with the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample
  • Have Cheminert (CX) termination and are designed to fit directly into the instrumentation removing the need for a needle
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 0.029 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 0.72 mm
Diameter (Metric) Outer 7.75 mm
Product Type Gastight Syringe
Max. Pressure 1000 psi
Max. Temperature (English) 239°F
Max. Temperature (Metric) 115°C
Min. Temperature (English) 50°F
Model 1702C
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 25 μL
For Use With (Equipment) Waters WISP HPLC Autosamplers