Learn More
Hamilton™ 800 Series Microliter™ Replacement Syringe Barrels
Product Code. 11572305
Click to view available options
For Use With (Equipment):
801 N Syringe
801 RN Syringe
802 RN Syringe
810 RN Syringe
8805 RN Syringe
8810 RN Syringe
8825 RN Syringe
Includes:
With needle (22s/51/2)
With needle (26s/51/2)
Without needle
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
Unit Size:
Each
Product Code. 11572305
Brand: Hamilton™ 32117
Barrels available with or without needle. Choose removable needle (RN) or cemented needle (N) termination. (See exact specifications below.)
Specifications
For Use With (Application) Liquids
Includes Without needle
Product Type Barrels/Plunger Assembly
Volume (Metric) 25 μL
For Use With (Equipment) 802 RN Syringe