Learn More
Hamilton™ Modified 7000 Microliter™ Syringes
Product Code. p-4117414
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
2 μL
5 μL
Needle Gauge:
22 G
22s G
23 G
24 G
25 G
Needle Point Number:
2
3
Needle Point Style:
Blunt
Sharp, Bevel, Curved, Non-coring
Unit Size:
Each
Product Code. Volume (Metric) Needle Gauge Needle Point Number Needle Point Style unitSize
10557361 0.5 μL 25 G 3 Blunt Each
10220812 0.5 μL 25 G 2 Sharp, Bevel, Curved, Non-coring Each
10008460 1 μL 22s G 3 Blunt Each
10399322 1 μL 22 G 2 Sharp, Bevel, Curved, Non-coring Each
10323761 2 μL 25 G 3 Blunt Each
10282011 2 μL 23 G 3 Blunt Each
10221921 5 μL 24 G 2 Sharp, Bevel, Curved, Non-coring Each
10401255 2 μL 23 G 2 Sharp, Bevel, Curved, Non-coring Each
10405952 2 μL 25 G 2 Sharp, Bevel, Curved, Non-coring Each
10791232 1 μL 25 G 3 Blunt Each
10332742 5 μL 24 G 3 Blunt Each
10028420 1 μL 25 G 2 Sharp, Bevel, Curved, Non-coring Each
12 options
Product Code. 10557361
Brand: Hamilton™ 86250
This series of syringes is ideal for dispensing volumes from 0.05 μL up to 5 μL. The 7000 series is the only zero dead volume syringe. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle.
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Autoclavable Autoclavable
Needle Gauge 25 G
Diameter (English) Inner 0.004 in.
Diameter (English) Inner Needle 0.004 in.
Diameter (Metric) Inner Needle 0.104 mm
Diameter (English) Outer Needle 0.02 in.
Diameter (English) Outer 0.3 in.
Diameter (Metric) Inner 0.1 mm
Diameter (Metric) Outer 7.82 mm
Diameter (Metric) Outer Needle 0.515 mm
Length (English) Needle 2.75 in.
Product Type General Purpose Syringe
Max. Pressure 6000 psi
Max. Temperature (English) 239°F
Max. Temperature (Metric) 115°C
Min. Temperature (English) 50°F
Needle Point Number 3
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 0.5 μL
Needle Point Style Blunt
For Use With (Equipment) HPLC Injection Valves
Connection Type Knurled Hub, Removable