Learn More
Hamilton™ Modified 7000 Microliter™ Syringes Product Code.: 10282011
  • ONLINE EXCLUSIVE PRICE
    1820.00 NOK
    1672.93 NOK / Each
    Save 147.07 NOK
    8% Off
Hamilton™ Modified 7000 Microliter™ Syringes
Product Code. 10282011
2 μL, 23 G, 3, Blunt, Each
Click to view available options
Volume (Metric):
0.5 μL
1 μL
2 μL
5 μL
Needle Gauge:
22 G
22s G
23 G
24 G
25 G
Needle Point Number:
2
3
Needle Point Style:
Blunt
Sharp, Bevel, Curved, Non-coring
Unit Size:
Each
Product Code. 10282011
Brand: Hamilton™ 88500
  • ONLINE EXCLUSIVE PRICE
    1820.00 NOK
    1672.93 NOK / Each
    Save 147.07 NOK
    8% Off
This series of syringes is ideal for dispensing volumes from 0.05 μL up to 5 μL. The 7000 series is the only zero dead volume syringe. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle.
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Autoclavable Autoclavable
Needle Gauge 23 G
Diameter (English) Inner 0.008 in.
Diameter (English) Inner Needle 0.008 in.
Diameter (Metric) Inner Needle 0.206 mm
Diameter (English) Outer Needle 0.02 in.
Diameter (English) Outer 0.3 in.
Diameter (Metric) Inner 0.2 mm
Diameter (Metric) Outer 7.82 mm
Diameter (Metric) Outer Needle 0.642 mm
Length (English) Needle 2.75 in.
Product Type General Purpose Syringe
Max. Pressure 6000 psi
Max. Temperature (English) 239°F
Max. Temperature (Metric) 115°C
Min. Temperature (English) 50°F
Needle Point Number 3
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 2 μL
Needle Point Style Blunt
For Use With (Equipment) HPLC Injection Valves
Connection Type Knurled Hub, Removable
Product Title
Hamilton™ Modified 7000 Microliter™ Syringes >