Learn More
Hamilton™ 1800 Series Gastight™ Replacement Syringe Barrels
Product Code. 15573963
Change view
Click to view available options
For Use With (Equipment):
1810 RN Syringe
Volume (Metric):
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Volume (Metric) Unit Size
15573963 1810 RN Syringe 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15573963 Supplier Hamilton™ Supplier No. 32190
  • ONLINE EXCLUSIVE PRICE
    1080.00 NOK
    867.92 NOK / Pack of 1
    Save 212.08 NOK
    19% Off
Estimated Shipment: 08-12-2026
Barrels range in volume from 10μ to 250μL with removable needle (RN) and cemented needle (N) terminations.
Specifications
Material PTFE
For Use With (Application) Gases and liquids
Product Type Replacement Syringe Barrel
Volume (Metric) 100 μL
For Use With (Equipment) 1810 RN Syringe