Learn More
Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: AD Termination
Product Code. 11557864
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
Max. Pressure:
1000 psi
200 psi
500 psi
700 psi
Model:
1001
1002
1005
1705
1710
1725
1750
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Max. Pressure Model unitSize
11557864 50 μL 1000 psi 1705 Each
10018422 100 μL 1000 psi 1710 Each
10268470 500 μL 700 psi 1750 Each
10440581 250 μL 500 psi 1725 Each
13586540 250 μL 500 psi 1725 Each
10688061 1 mL 200 psi 1001 Each
10698061 2.5 mL 200 psi 1002 Each
10186560 5 mL 200 psi 1005 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11557864 Supplier Hamilton™ Supplier No. 201000
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • AccuDil™ (AD) - This termination is designed for MICROLAB 1000 Diluters/Dispensers
Specifications
Material Borosilicate Glass (Barrel), PTFE (Tip)
Product Type Gastight Syringe
Max. Pressure 1000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 68°F
Model 1705
Needle Point Number No Needle Included
Quantity Ea.
Volume (Metric) 50 μL
Product Line AccuDil
For Use With (Equipment) Microlab 1000 Diluters/Dispensers