Learn More
O&M Halyard Quick Check Sterilization Wrap (H300)
GREENER_CHOICE
Product Code. 30274698 Shop All Halyard Products
Change view
Click to view available options
Dimensions (L x W):
101 x 101 cm (40 x 40 in.)
114 x 114 cm (45 x 45 in.)
121 x 121 cm (48 x 48 in.)
137 x 137 cm (54 x 54 in.)
137 x 182 cm (54 x 72 in.)
60 x 60 cm (24 x 24 in.)
76 x 76 cm (30 x 30 in.)
91 x 91 cm (36 x 36 in.)
Unit Size:
Pack of 144
Pack of 240
Pack of 48
Pack of 96
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
30274698 76 x 76 cm (30 x 30 in.) Pack of 144
30274770 60 x 60 cm (24 x 24 in.) Pack of 240
30274757 91 x 91 cm (36 x 36 in.) Pack of 144
30274743 101 x 101 cm (40 x 40 in.) Pack of 96
30274724 114 x 114 cm (45 x 45 in.) Pack of 48
30274741 121 x 121 cm (48 x 48 in.) Pack of 96
30274778 137 x 137 cm (54 x 54 in.) Pack of 48
30274768 137 x 182 cm (54 x 72 in.) Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 30274698 Supplier O&M Supplier No. 34173
Using Halyard Quick Check wraps is a fast, easy way to ensure instrument sterility is uncompromised.
  • Inspect trays easier and faster than ever
  • Superior microbial barrier protection with patented Powerguard technology
  • Cleared for all of the same sterilization systems as Halyard Kimguard One-step Wrap
  • Made from recyclable No. 5 polypropylene fabric
  • Made in the USA
Specifications
Autoclavable Autoclavable
Certifications/Compliance Meets NFPA Class 1
Color Blue
For Use With (Equipment) Basin sets, instrument sets, procedure trays and large linen packs (6 to 9 lb.)
Material SMS Fabric (H300)
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 76 x 76 cm (30 x 30 in.)
Length (English) 30 in.
Width (English) 30 in.
Product Type Sterilization Wrap
Length (Metric) 76 cm
Width (Metric) 76 cm