Learn More
Applied Biosystems™ Halogen Lamp for Applied Biosystems Real-Time PCR Systems
Product Code. 15820458 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820458 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15820458 Supplier Applied Biosystems™ Supplier No. 100052500
Estimated Shipment: 17-11-2026
This halogen lamp is an accessory for the 7300, 7500, 7500 Fast, ViiA7, QuantStudio 6 Flex and QuantStudio 7 Flex Systems.
  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY
Specifications
Light Source Halogen
Content And Storage Store at room temperature
For Use With (Equipment) 7300, 7500, 7500 Fast, ViiA7, QuantStudio 6 Flex, QuantStudio 7 Flex Systems
Product Type Halogen Lamp
Quantity 1 each
For Research Use Only. Not for use in diagnostics procedures.