Description
- Halogen bulb
- 15 V / 150 W
- For use with KL1500 LCD / KL1500 HAL and KL1500 compact
Order Info
Voltage
Specifications
Specifications
| Type | Replacement Bulb |
| Bulb Type | Halogen |
| For Use With (Equipment) | Cold Light Source KL1500HAL |
| Quantity | 1 Ea. |
| Voltage | 15 V |
| Wattage | 150 w |