Learn More
Macherey-Nagel™ Haloform Test Mixture in n-Pentane
Product Code. 10216421 Shop All Macherey-Nagel Products
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10216421 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10216421 Supplier Macherey-Nagel™ Supplier No. 722311
  • ONLINE EXCLUSIVE PRICE
    932.00 NOK
    818.34 NOK / 1mL
    Save 113.66 NOK
    12% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Specifications
For Use With (Application) For GC Environmental Analyses
Includes Dichloromethane (795 ng/mL), Trichloromethane (75 ng/mL), 1,1,1-Trichloroethane (67 ng/mL), Tetrachloromethane (80 ng/mL), Trichloroethene (73 ng/mL), Bromodichloromethane (100 ng/mL), Dibromochloromethane (122 ng/mL), Tetrachloroethene (81 ng/mL), Tribromomethane (145 ng/mL)
Volume (Metric) 1 mL
Product Type Haloform Test Mixture in n-pentane