Learn More
3M™ Half Mask Respirator Kits
Product Code. 15352092 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15352092 Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15352092 Supplier 3M™ Supplier No. 7000061547
  • ONLINE EXCLUSIVE PRICE
    1020.00 NOK
    671.68 NOK / Pack of 1
    Save 348.32 NOK
    34% Off
View all promotions
Estimated Shipment: 17-11-2026
Specifications
Cartridge Type A12P R
Certifications/Compliance CE Approved
Color Gray
Product Type Half Mask Respirator Kit
Size Medium
For Use With (Application) Agriculture,‎ Construction,‎ Paint spraying
Includes Reusable half face mask and set of filters to protect against organic vapors and particulates
Model 6200
Quantity 1/Pk.