missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25940-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.Specifications
HAL | |
Polyclonal | |
Unconjugated | |
HAL | |
HAL, HIS, Histidase, histidine ammonia lyase, HSTD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15109, 29301, 3034 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P21213, P35492, P42357 | |
Hal | |
HAL Fusion Protein Ag22985 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |