missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27593-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.Specifications
HAI-1 | |
Polyclonal | |
Unconjugated | |
SPINT1 | |
HAI, HAI 1, HAI1, MANSC2, SPINT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6692 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43278 | |
SPINT1 | |
HAI-1 Fusion Protein Ag26733 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |