Learn More
Hecht Karl™ Haemacytometer Coverslips
Product Code. 10043601
Change view
Click to view available options
Width (Metric):
22 mm
Unit Size:
Pack of 10
Product Code. Width (Metric) unitSize
10043601 22 mm Pack of 10
1 options
Product Code. 10043601
Brand: Hecht Karl™ 40416710
Only null left
Specifications
Length (Metric) 22 mm
Width (Metric) 22 mm
Thickness (Metric) 0.4 mm
Dimensions (L x W) 22 x 22 mm